Butylbenzoat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzoic acid butyl ester
A17010.0B
A17010.30
A17010.0BEA
80.7
EUR
A17010.0B
A17010.30
Butylbenzoat 98%
Butylbenzoat
Purity: 98%
CAS No.: 136-60-7
Molecular Formula: C11H14O2
Formula Weight: 178.23
Flash Point: 106°(222°F)
Refractive Index: 1.4980
Melting Point: -22°
Boiling Point: 250°
Beilstein Reference: 1867073
Merck Reference: 14,1552
MDL No.: MFCD00009439
Formel:
C₁₁H₁₄O₂ Molecular Weight: 178,23 g/mol Siedepunkt: 250 °C Schmelzpunkt: –22 °C Dichte: 1,011 Flammpunkt: 106 °C (222 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009439 CAS-Nummer: 136-60-7 EINECS: 205-252-7 Merck Index: 14,1552 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...