Ethylbenzoat ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Benzoic acid ethyl ester

A13057.0E A13057.22 A13057.36
A13057.0EEA 134 EUR
A13057.0E A13057.22 A13057.36
Ethylbenzoat ≥99%
Ethylbenzoat

Purity: 99% CAS No.: 93-89-0 Molecular Formula: C9H10O2 Formula Weight: 150.18 Flash Point: 84°(183°F) Refractive Index: 1.5050 Melting Point: -34° Boiling Point: 212-214° Beilstein Reference: 1908172 Merck Reference: 14,3766 MDL No.: MFCD00009109

Formel: C₉H₁₀O₂
Molecular Weight: 150,18 g/mol
Siedepunkt: 212…214 °C
Schmelzpunkt: –34 °C
Dichte: 1,049
Flammpunkt: 84 °C (183 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009109
CAS-Nummer: 93-89-0
EINECS: 202-284-3
Merck Index: 14,3766

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR