Hexylbenzoat ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Benzoic acid hexyl ester
L06293.14
L06293.22
L06293.14EA
30.1
EUR
L06293.14
L06293.22
Hexylbenzoat ≥98%
Hexylbenzoat
Purity: 98+%
CAS No.: 6789-88-4
Molecular Formula: C13H18O2
Formula Weight: 206.29
Flash Point: 148°(298°F)
Refractive Index: 1.4930
Boiling Point: 272°
Beilstein Reference: 2048117
MDL No.: MFCD00051714
Formel:
C₁₃H₁₈O₂ Molecular Weight: 206,28 g/mol Siedepunkt: 272 °C Dichte: 0,980 Flammpunkt: 148 °C (298 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051714 CAS-Nummer: 6789-88-4 EINECS: 229-856-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...