3-Cyanbenzoesäure

Lieferant: FLUOROCHEM
Achtung

013809-25G 013809-100G
FLUO013809-25GEA 0 EUR
FLUO013809-25G FLUO013809-100G
3-Cyanbenzoesäure
3-Cyanbenzoesäure
Formel: C₈H₅NO₂
Molecular Weight: 147.13 g/mol
Schmelzpunkt: 219
Lagertemperatur: Raumtemperatur
MDL: MFCD00002486
CAS-Nummer: 1877-72-1
EINECS: 217-511-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR