5-Nitroanthranilsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-5-nitrobenzoic acid

245950050 245950250
ACRO245950050EA 32.1 EUR
ACRO245950050 ACRO245950250
5-Nitroanthranilsäure 98%
5-Nitroanthranilsäure
Formel: C₇H₆N₂O₄
Molecular Weight: 182,14 g/mol
Schmelzpunkt: 278 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017039
CAS-Nummer: 616-79-5
EINECS: 210-493-6

Bestellen

Spezifikation Testergebnisse

Appearance Bright yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR