5-Nitroanthranilsäure

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-5-nitrobenzoic acid

BTB07484.10 BTB07484.50
MAYBBTB07484.10EA 0 EUR
MAYBBTB07484.10 MAYBBTB07484.50
5-Nitroanthranilsäure
5-Nitroanthranilsäure
Formel: C₇H₆N₂O₄
Molecular Weight: 182,14 g/mol
Schmelzpunkt: 278 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017039
CAS-Nummer: 616-79-5
EINECS: 210-493-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR