5-Amino-2-nitrobenzoesäure 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Nitro-5-aminobenzoic acid, NSC 74455, 5-Amino-2-nitrobenzoic Acid, 3-Carboxy-4-nitroaniline
L16225.06
L16225.14
L16225.06EA
81.3
EUR
L16225.06
L16225.14
5-Amino-2-nitrobenzoesäure 95%
5-Amino-2-nitrobenzoesäure
Purity: 95%
CAS No.: 13280-60-9
Molecular Formula: C7H6N2O4
Formula Weight: 182.14
Melting Point: ca 240° dec.
Beilstein Reference: 2107512
Formel:
C₇H₆N₂O₄ Molecular Weight: 182,14 g/mol Siedepunkt: 481 °C (1013 hPa) Schmelzpunkt: ∼240 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00035748 CAS-Nummer: 13280-60-9 EINECS: 236-283-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...