3,4,5-Trimethoxyanthranilsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

A14042.06 A14042.14
A14042.06EA 96.9 EUR
A14042.06 A14042.14
3,4,5-Trimethoxyanthranilsäure 97%
3,4,5-Trimethoxyanthranilsäure

Purity: 97% CAS No.: 61948-85-4 Molecular Formula: C10H13NO5 Formula Weight: 227.22 Melting Point: 137-141° Beilstein Reference: 3353856

Formel: C₁₀H₁₃NO₅
Molecular Weight: 227.22 g/mol
Schmelzpunkt: 137…141 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051630
CAS-Nummer: 61948-85-4
EINECS: 263-344-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR