5-Hydroxyanthranilsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Hydroxyanthranilic acid
L08256.14
L08256.06
L08256.14EA
139
EUR
L08256.14
L08256.06
5-Hydroxyanthranilsäure 98%
5-Hydroxyanthranilsäure
Purity: 98%
CAS No.: 394-31-0
Molecular Formula: C7H7NO3
Formula Weight: 153.14
Melting Point: ca 248° dec.
Beilstein Reference: 2803663
Formel:
C₇H₇NO₃ Molecular Weight: 153,14 g/mol Siedepunkt: 347 °C (1013 hPa) Schmelzpunkt: ∼248 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007870 CAS-Nummer: 394-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...