4-Amino-3-hydroxybenzoesäure 98%

Lieferant: Thermo Scientific
Achtung

354260050 354260250
ACRO354260050EA 43.8 EUR
ACRO354260050 ACRO354260250
4-Amino-3-hydroxybenzoesäure 98%
4-Amino-3-hydroxybenzoic acid
Formel: C₇H₇NO₃
Molecular Weight: 153,14 g/mol
Schmelzpunkt: 211…215 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017094
CAS-Nummer: 2374-03-0

Bestellen

Spezifikation Testergebnisse

Appearance Yellow-brown to brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR