3,4-Diaminobenzoesäure 94%
Lieferant: Thermo Scientific
|
Achtung
|
A14213.14
A14213.22
A14213.36
A14213.14EA
46.9
EUR
A14213.14
A14213.22
A14213.36
3,4-Diaminobenzoesäure 94%
3,4-Diaminobenzoic acid
Purity: 94%
CAS No.: 619-05-6
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: ca 210° dec.
Beilstein Reference: 775892
Formel:
C₇H₈N₂O₂ Molecular Weight: 152,15 g/mol Siedepunkt: 354 °C (1013 hPa) Schmelzpunkt: ∼210 °C (Zers.) Dichte: 1,544 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007726 CAS-Nummer: 619-05-6 EINECS: 210-577-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...