4-Nitro-o-anissäure

Lieferant: CHEMPUR
Achtung

FL12257.POR 012257-5G
CHMPFL12257.POREA 0 EUR
CHMPFL12257.POR FLUO012257-5G
4-Nitro-o-anissäure
4-Nitro-o-anissäure
Formel: C₈H₇NO₅
Molecular Weight: 197,15 g/mol
MDL: MFCD00216573
CAS-Nummer: 2597-56-0
EINECS: 219-998-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR