6-Nitro-m-anissäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Methoxy-2-nitrobenzoic acid

L13346.06 L13346.14
L13346.06EA 20.9 EUR
L13346.06 L13346.14
6-Nitro-m-anissäure ≥97%
6-Nitro-m-anissäure

Purity: 97% CAS No.: 1882-69-5 Molecular Formula: C8H7NO5 Formula Weight: 197.15 Melting Point: 130-135° Beilstein Reference: 1877657

Formel: C₈H₇NO₅
Molecular Weight: 197,15 g/mol
Schmelzpunkt: 130…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00151836
CAS-Nummer: 1882-69-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR