6-Nitro-m-anissäure
Lieferant: CHEMPUR
Synonyme:
5-Methoxy-2-nitrobenzoic acid
RH00891.250
RH00891.1
CHMPRH00891.250EA
0
EUR
CHMPRH00891.250
MAYBRH00891.1
6-Nitro-m-anissäure
6-Nitro-m-anissäure
Formel:
C₈H₇NO₅ Molecular Weight: 197,15 g/mol Schmelzpunkt: 138 °C |
MDL:
MFCD00151836 CAS-Nummer: 1882-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...