4-Hydroxy-3-nitrobenzoesäure
Lieferant: Thermo Scientific
|
Achtung
|
BTB10601.1
BTB10601.10
BTB10601.50
MAYBBTB10601.1EA
0
EUR
MAYBBTB10601.1
MAYBBTB10601.10
MAYBBTB10601.50
4-Hydroxy-3-nitrobenzoesäure
4-Hydroxy-3-nitrobenzoic acid
Formel:
C₇H₅NO₅ Molecular Weight: 183,12 g/mol Schmelzpunkt: 182…185 °C Dichte: 1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007119 CAS-Nummer: 616-82-0 EINECS: 210-494-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...