2,3-Dimethoxybenzoesäure ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
A14592.09
A14592.18
A14592.09EA
28.8
EUR
A14592.09
A14592.18
2,3-Dimethoxybenzoesäure ≥99%
2,3-Dimethoxybenzoic acid
Purity: 99%
CAS No.: 1521-38-6
Molecular Formula: C9H10O4
Formula Weight: 182.18
Odour: Odourless
Melting Point: 121-124°
Beilstein Reference: 2210858
MDL No.: MFCD00002432
Formel:
C₉H₁₀O₄ Molecular Weight: 182,18 g/mol Siedepunkt: 314 °C (1013 hPa) Schmelzpunkt: 121…124 °C Dichte: 1,387 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002432 CAS-Nummer: 1521-38-6 EINECS: 216-188-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...