3-Hydroxybenzoesäure ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
m-Hydroxybenzoic Acid
A13628.22
A13628.36
A13628.22EA
37.9
EUR
A13628.22
A13628.36
3-Hydroxybenzoesäure ≥99%
3-Hydroxybenzoesäure
Purity: 99%
CAS No.: 99-06-9
Molecular Formula: C7H6O3
Formula Weight: 138.12
Flash Point: 220°(428°F)
Odour: Odourless
Beilstein Reference: 508160
MDL No.: MFCD00002506
Formel:
C₇H₆O₃ Molecular Weight: 138,12 g/mol Siedepunkt: 346 °C (1013 hPa) Schmelzpunkt: 201…203 °C Dichte: 1,473 g/cm³ (20 °C) Flammpunkt: 220 °C (428 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002506 CAS-Nummer: 99-06-9 EINECS: 202-726-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...