3,5-Dihydroxybenzoesäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
alpha-Resorcylic acid
114901000
114905000
ACRO114901000EA
29.4
EUR
ACRO114901000
ACRO114905000
3,5-Dihydroxybenzoesäure 97%
3,5-Dihydroxybenzoic acid
Formel:
C₇H₆O₄ Molecular Weight: 154,12 g/mol Schmelzpunkt: 235…240 °C Flammpunkt: >200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002512 CAS-Nummer: 99-10-5 EINECS: 202-730-7 |
Spezifikation Testergebnisse
Appearance | White to beige to light pink Powder |
Infrared spectrum | Conforms |
Melting point | 235 to 242 °C |
HPLC | ≥96.0 % |
Titration with NaOH | 96.0 to 104.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...