2-Phenoxybenzoesäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Carboxydiphenylether
A10515.14
A10515.22
A10515.14EA
61.5
EUR
A10515.14
A10515.22
2-Phenoxybenzoesäure ≥98%
2-Phenoxybenzoesäure
Purity: 98%
CAS No.: 2243-42-7
Molecular Formula: C13H10O3
Formula Weight: 214.22
Melting Point: 111-115°
Beilstein Reference: 978452
Formel:
C₁₃H₁₀O₃ Molecular Weight: 214,22 g/mol Siedepunkt: 355 °C (1013 hPa) Schmelzpunkt: 111…115 °C Dichte: 1,2141 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002429 CAS-Nummer: 2243-42-7 EINECS: 218-811-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...