4-Phenoxybenzoesäure 98%

Lieferant: Thermo Scientific
Achtung

352720010 352720050 352720250
ACRO352720010EA 17.5 EUR
ACRO352720010 ACRO352720050 ACRO352720250
4-Phenoxybenzoesäure 98%
4-Phenoxybenzoesäure
Formel: C₁₃H₁₀O₃
Molecular Weight: 214,22 g/mol
Schmelzpunkt: 160…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002539
CAS-Nummer: 2215-77-2
EINECS: 218-682-5

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 160 to 164 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR