1,2,4-Trichlorbenzol ≥99%
Lieferant: Thermo Scientific
|
Achtung
|
A10130.36
A10130.36EA
34.1
EUR
A10130.36
1,2,4-Trichlorbenzol ≥99%
1,2,4-Trichlorobenzene
Purity: 99%
CAS No.: 120-82-1
Molecular Formula: C6H3Cl3
Formula Weight: 181.45
Flash Point: 110°(230°F)
Refractive Index: 1.572
Melting Point: 17°
Boiling Point: 213°
Solubility: Miscible with ether, benzene, CS2
Merck Reference: 14,9631
UN No.: 2321
MDL No.: MFCD00000547
Formel:
C₆H₃Cl₃ Molecular Weight: 181,45 g/mol Siedepunkt: 213 °C Schmelzpunkt: 17 °C Dichte: 1,454 Flammpunkt: 110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000547 CAS-Nummer: 120-82-1 EINECS: 204-428-0 UN: 2321 ADR: 6.1,III Merck Index: 14,9631 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...