3-Chlorbenzaldehyd ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
A13254.14
A13254.22
A13254.36
A13254.14EA
38.6
EUR
A13254.14
A13254.22
A13254.36
3-Chlorbenzaldehyd ≥97%
3-Chlorbenzaldehyd
Purity: 97%
CAS No.: 587-04-2
Molecular Formula: C7H5ClO
Formula Weight: 140.57
Flash Point: 88°(190°F)
Refractive Index: 1.5640
Odour: Characteristic
Melting Point: 16-18°
Boiling Point: 213-214°
Beilstein Reference: 507098
MDL No.: MFCD00003350
Formel:
C₇H₅ClO Molecular Weight: 140,57 g/mol Siedepunkt: 213…214 °C Schmelzpunkt: 16…18 °C Dichte: 1,239 Flammpunkt: 88 °C (190 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003350 CAS-Nummer: 587-04-2 EINECS: 209-596-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...