2,4-Dichlorbenzonitril
Lieferant: Thermo Scientific
TL00187DA
TL00187EA
TL00187.50
MAYBTL00187DAEA
0
EUR
MAYBTL00187DA
MAYBTL00187EA
MAYBTL00187.50
2,4-Dichlorbenzonitril
2,4-Dichlorbenzonitril
Formel:
C₇H₃Cl₂N Molecular Weight: 172,01 g/mol Siedepunkt: 261 °C (1013 hPa) Schmelzpunkt: 52…53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016373 CAS-Nummer: 6574-98-7 EINECS: 229-493-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...