2,5-Dichlorbenzonitril

Lieferant: FLUOROCHEM

018514-1G 018514-5G
FLUO018514-1GEA 50.3 EUR
FLUO018514-1G FLUO018514-5G
2,5-Dichlorbenzonitril
2,5-Dichlorbenzonitril
Formel: C₇H₃Cl₂N
Molecular Weight: 172.01 g/mol
Siedepunkt: 269
Schmelzpunkt: 130…132 °C
MDL: MFCD00019741
CAS-Nummer: 21663-61-6
EINECS: 244-518-7
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR