3,5-Dichlorbenzonitril

Lieferant: FLUOROCHEM

018516-1G 018516-5G
FLUO018516-1GEA 31.1 EUR
FLUO018516-1G FLUO018516-5G
3,5-Dichlorbenzonitril
3,5-Dichlorbenzonitril
Formel: C₇H₃Cl₂N
Molecular Weight: 172,01 g/mol
Siedepunkt: 269 °C (1013 hPa)
Schmelzpunkt: 66 °C
MDL: MFCD00001800
CAS-Nummer: 6575-00-4
EINECS: 229-495-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR