2-Amino-6-chlorbenzonitril 98%

Lieferant: Thermo Scientific
Gefahr

L06883.03 L06883.06
L06883.03EA 25.2 EUR
L06883.03 L06883.06
2-Amino-6-chlorbenzonitril 98%
2-Amino-6-chlorbenzonitril

Purity: 98% CAS No.: 6575-11-7 Molecular Formula: C7H5ClN2 Formula Weight: 152.58 Melting Point: 133-137° Beilstein Reference: 2717090 UN No.: 3439 MDL No.: MFCD00015546

Formel: C₇H₅ClN₂
Molecular Weight: 152,58 g/mol
Siedepunkt: 301 °C (1013 hPa)
Schmelzpunkt: 133…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015546
CAS-Nummer: 6575-11-7
EINECS: 229-501-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR