4-Amino-2-chlorbenzonitril ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-Chlor-4-cyananilin, 4-Amino-2-chlorbenzoesäurenitril

L15924.06 L15924.14 L15924.22
L15924.06EA 26.2 EUR
L15924.06 L15924.14 L15924.22
4-Amino-2-chlorbenzonitril ≥98%
4-Amino-2-chlorbenzonitril

Purity: 98+% CAS No.: 20925-27-3 Molecular Formula: C7H5ClN2 Formula Weight: 152.58 Melting Point: 116-119° Beilstein Reference: 2936425 UN No.: 3439 MDL No.: MFCD00035926

Formel: C₇H₅ClN₂
Molecular Weight: 152,58 g/mol
Siedepunkt: 301 °C (1013 hPa)
Schmelzpunkt: 116…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00035926
CAS-Nummer: 20925-27-3
EINECS: 244-114-0
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR