4-Chlor-3-nitrobenzonitril ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
A12648.06
A12648.14
A12648.22
A12648.06EA
42.6
EUR
A12648.06
A12648.14
A12648.22
4-Chlor-3-nitrobenzonitril ≥98%
4-Chlor-3-nitrobenzonitril
Purity: 98%
CAS No.: 939-80-0
Molecular Formula: C7H3ClN2O2
Formula Weight: 182.57
Melting Point: 98-102°
Beilstein Reference: 1639111
Formel:
C₇H₃ClN₂O₂ Molecular Weight: 182,57 g/mol Schmelzpunkt: 98…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016987 CAS-Nummer: 939-80-0 EINECS: 213-364-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...