2,3-Dichlorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

001164-5G 001164-25G
FLUO001164-5GEA 0 EUR
FLUO001164-5G FLUO001164-25G
2,3-Dichlorbenzoesäure
2,3-Dichlorobenzoic acid
Formel: C₇H₄Cl₂O₂
Molecular Weight: 191,01 g/mol
Schmelzpunkt: 168,3 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002413
CAS-Nummer: 50-45-3
EINECS: 200-039-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR