2,6-Dichlorbenzoesäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A10542.06 A10542.14 A10542.22
A10542.06EA 16.2 EUR
A10542.06 A10542.14 A10542.22
2,6-Dichlorbenzoesäure ≥98%
2,6-Dichlorobenzoic acid

Purity: 98% CAS No.: 50-30-6 Molecular Formula: C7H4Cl2O2 Formula Weight: 191.01 Melting Point: 141-145° Beilstein Reference: 973858

Formel: C₇H₄Cl₂O₂
Molecular Weight: 191.01 g/mol
Siedepunkt: 299
Schmelzpunkt: 141…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002418
CAS-Nummer: 50-30-6
EINECS: 200-025-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR