3,5-Dichloranthranilsäure

Lieferant: Thermo Scientific

Synonyme: 2-Amino-3,5-dichlorobenzoic acid

RDP00048.1 RDP00048.10 RDP00048.50
MAYBRDP00048.1EA 0 EUR
MAYBRDP00048.1 MAYBRDP00048.10 MAYBRDP00048.50
3,5-Dichloranthranilsäure
3,5-Dichloranthranilsäure
Formel: C₇H₅Cl₂NO₂
Molecular Weight: 206,03 g/mol
Siedepunkt: 344 °C (1013 hPa)
Schmelzpunkt: 226…229 °C
MDL: MFCD00017088
CAS-Nummer: 2789-92-6
EINECS: 220-520-3
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR