5-Chloranthranilsäure 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-5-chlorobenzoic acid

A14329.06 A14329.14 A14329.22
A14329.06EA 37.4 EUR
A14329.06 A14329.14 A14329.22
5-Chloranthranilsäure 98%
5-Chloranthranilsäure

Purity: 98% CAS No.: 635-21-2 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: ca 208° dec. Beilstein Reference: 639027

Formel: C₇H₆ClNO₂
Molecular Weight: 171,58 g/mol
Schmelzpunkt: ∼ 208 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007838
CAS-Nummer: 635-21-2
EINECS: 211-230-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR