5-Chloranthranilsäure

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Amino-5-chlorobenzoic acid

RDP00097.1 RDP00097.10 RDP00097.50
MAYBRDP00097.1EA 0 EUR
MAYBRDP00097.1 MAYBRDP00097.10 MAYBRDP00097.50
5-Chloranthranilsäure
5-Chloranthranilsäure
Formel: C₇H₆ClNO₂
Molecular Weight: 171,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007838
CAS-Nummer: 635-21-2
EINECS: 211-230-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR