5-Chloranthranilsäure

Lieferant: FLUOROCHEM
Achtung

Synonyme: 2-Amino-5-chlorobenzoic acid

011364-100G 011364-25G
FLUO011364-100GEA 130 EUR
FLUO011364-100G FLUO011364-25G
5-Chloranthranilsäure
5-Chloranthranilsäure
Formel: C₇H₆ClNO₂
Molecular Weight: 171,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007838
CAS-Nummer: 635-21-2
EINECS: 211-230-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR