4-Chlor-1,2-difluorbenzol

Lieferant: Thermo Scientific

Synonyme: 3,4-Difluorochlorobenzene

SB01824.1 SB01824EA SB01824.50
MAYBSB01824.1EA 0 EUR
MAYBSB01824.1 MAYBSB01824EA MAYBSB01824.50
4-Chlor-1,2-difluorbenzol
4-Chlor-1,2-difluorbenzol
Formel: C₆H₃ClF₂
Molecular Weight: 148.54 g/mol
Siedepunkt: 118…120 °C (1013 hPa)
Dichte: 1.35 g/cm³ (20 °C)
MDL: MFCD00042572
CAS-Nummer: 696-02-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR