2-Chlor-4-fluorbenzaldehyd ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
A15345.03
A15345.06
A15345.03EA
14.6
EUR
A15345.03
A15345.06
2-Chlor-4-fluorbenzaldehyd ≥97%
2-Chloro-4-fluorobenzaldehyde
Purity: 97%
CAS No.: 84194-36-5
Molecular Formula: C7H4ClFO
Formula Weight: 158.56
Flash Point:
Formel:
C₇H₄ClFO Molecular Weight: 158.56 g/mol Siedepunkt: 118…120 °C (50 mmHg) Schmelzpunkt: 60…63 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042527 CAS-Nummer: 84194-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...