2-Chlor-6-fluorbenzaldehyd ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

A10513.14 A10513.22
A10513.14EA 29 EUR
A10513.14 A10513.22
2-Chlor-6-fluorbenzaldehyd ≥97%
2-Chloro-6-fluorobenzaldehyde

Purity: 97% CAS No.: 387-45-1 Molecular Formula: C7H4ClFO Formula Weight: 158.56 Flash Point: 101°(213°F) Melting Point: 35-39° Boiling Point: 91-93°/10mm Beilstein Reference: 2245530 MDL No.: MFCD00003306

Formel: C₇H₄ClFO
Molecular Weight: 158.56 g/mol
Siedepunkt: 91…93 °C (10 mmHg)
Schmelzpunkt: 35…39 °C
Flammpunkt: 101 °C (213 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00003306
CAS-Nummer: 387-45-1
EINECS: 206-860-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR