2-Chlor-4-fluorbenzonitril ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
A15478.06
A15478.14
A15478.06EA
51.8
EUR
A15478.06
A15478.14
2-Chlor-4-fluorbenzonitril ≥98%
2-Chlor-4-fluorbenzonitril
Purity: 98%
CAS No.: 60702-69-4
Molecular Formula: C7H3ClFN
Formula Weight: 155.56
Melting Point: 62-66°
Beilstein Reference: 6323034
UN No.: 3439
MDL No.: MFCD00042523
Formel:
C₇H₃ClFN Molecular Weight: 155,56 g/mol Siedepunkt: 237 °C (1013 hPa) Schmelzpunkt: 62…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042523 CAS-Nummer: 60702-69-4 EINECS: 262-384-8 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...