2-Chlor-4-fluorbenzonitril

Lieferant: FLUOROCHEM

004432-25G 004432-100G
FLUO004432-25GEA 57.9 EUR
FLUO004432-25G FLUO004432-100G
2-Chlor-4-fluorbenzonitril
2-Chlor-4-fluorbenzonitril
Formel: C₇H₃ClFN
Molecular Weight: 155,56 g/mol
Siedepunkt: 237 °C (1013 hPa)
Schmelzpunkt: 63…65 °C
MDL: MFCD00042523
CAS-Nummer: 60702-69-4
EINECS: 262-384-8
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR