4-Chlor-2-fluorbenzonitril ≥98%

Lieferant: Thermo Scientific
Achtung

L20160.03 L20160.06 L20160.14
L20160.03EA 19.1 EUR
L20160.03 L20160.06 L20160.14
4-Chlor-2-fluorbenzonitril ≥98%
4-Chlor-2-fluorbenzonitril

Purity: 98+% CAS No.: 57381-51-8 Molecular Formula: C7H3ClFN Formula Weight: 155.56 Melting Point: 58-61° Beilstein Reference: 4231142 UN No.: 3439 MDL No.: MFCD00143284

Formel: C₇H₃ClFN
Molecular Weight: 155,56 g/mol
Siedepunkt: 237 °C (1013 hPa)
Schmelzpunkt: 58…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143284
CAS-Nummer: 57381-51-8
EINECS: 260-712-4
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR