2,4-Dichlor-5-fluorbenzoesäure

Lieferant: FLUOROCHEM
Achtung

004710-5G 004710-1G
FLUO004710-5GEA 198 EUR
FLUO004710-5G FLUO004710-1G
2,4-Dichlor-5-fluorbenzoesäure
2,4-Dichloro-5-fluorobenzoic acid
Formel: C₇H₃Cl₂FO₂
Molecular Weight: 209 g/mol
Siedepunkt: 298
Schmelzpunkt: 141…144 °C
MDL: MFCD00075281
CAS-Nummer: 86522-89-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR