2-Chlor-4-fluorbenzoesäure

Lieferant: FLUOROCHEM

CFF930. 004428-25G 004428-100G
CHMPCFF930.EA 0 EUR
CHMPCFF930. FLUO004428-25G FLUO004428-100G
2-Chlor-4-fluorbenzoesäure
2-Chloro-4-fluorobenzoic acid
Formel: C₇H₄ClFO₂
Molecular Weight: 174.56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00010615
CAS-Nummer: 2252-51-9
EINECS: 218-845-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR