3-Chlor-4-fluorbenzoesäure
Lieferant: Thermo Scientific
BTB03715.1
BTB03715.10
MAYBBTB03715.1EA
0
EUR
MAYBBTB03715.1
MAYBBTB03715.10
3-Chlor-4-fluorbenzoesäure
3-Chloro-4-fluorobenzoic acid
Formel:
C₇H₄ClFO₂ Molecular Weight: 174.56 g/mol Siedepunkt: 273 Schmelzpunkt: 136 |
MDL:
MFCD00042477 CAS-Nummer: 403-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...