3-Chlor-4-fluorbenzoesäure

Lieferant: FLUOROCHEM

CFF933. 002149.5G 002149-25G 002149-100G
CHMPCFF933.EA 0 EUR
CHMPCFF933. FLUO002149.5G FLUO002149-25G FLUO002149-100G
3-Chlor-4-fluorbenzoesäure
3-Chloro-4-fluorobenzoic acid
Formel: C₇H₄ClFO₂
Molecular Weight: 174.56 g/mol
Siedepunkt: 273
Schmelzpunkt: 136
MDL: MFCD00042477
CAS-Nummer: 403-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR