4-Chlor-2-fluorbenzoesäure
Lieferant: CHEMPUR
|
Achtung
|
BTB13066.250
BTB13066.1
CHMPBTB13066.250EA
0
EUR
CHMPBTB13066.250
MAYBBTB13066.1
4-Chlor-2-fluorbenzoesäure
4-Chloro-2-fluorobenzoic acid
Formel:
C₇H₄ClFO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042468 CAS-Nummer: 446-30-0 EINECS: 207-162-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...