3-Chlor-2-fluoranilin

Lieferant: FLUOROCHEM

CFF917. 004418-25G 004418-100G
CHMPCFF917.EA 0 EUR
CHMPCFF917. FLUO004418-25G FLUO004418-100G
3-Chlor-2-fluoranilin
3-Chlor-2-fluoranilin
Formel: C₆H₅ClFN
Molecular Weight: 145.56 g/mol
Siedepunkt: 99 °C (14 mmHg)
MDL: MFCD00069415
CAS-Nummer: 2106-04-9
EINECS: 218-283-6
UN: 2810
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR