5-Chlor-2-fluoranilin
Lieferant: Thermo Scientific
|
Achtung
|
SB01826.1
SB01826.10
MAYBSB01826.1EA
0
EUR
MAYBSB01826.1
MAYBSB01826.10
5-Chlor-2-fluoranilin
5-Chlor-2-fluoranilin
Formel:
C₆H₅ClFN Molecular Weight: 145,56 g/mol |
MDL:
MFCD00069416 CAS-Nummer: 2106-05-0 EINECS: 218-284-1 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...