4-Chlor-2-fluoranilin ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
A16131.14
A16131.22
A16131.14EA
75
EUR
A16131.14
A16131.22
4-Chlor-2-fluoranilin ≥98%
4-Chlor-2-fluoranilin
Purity: 98%
CAS No.: 57946-56-2
Molecular Formula: C6H5ClFN
Formula Weight: 145.56
Flash Point: 99°(210°F)
Refractive Index: 1.5600
Boiling Point: 92°/2mm
Beilstein Reference: 1934804
UN No.: 2810
MDL No.: MFCD00010625
Formel:
C₆H₅ClFN Siedepunkt: 92 °C (2 mmHg) Dichte: 1,311 Flammpunkt: 99 °C (210 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010625 CAS-Nummer: 57946-56-2 EINECS: 261-034-1 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...