4-Chlor-3-fluoranilin

Lieferant: FLUOROCHEM

CFF919. 009745-1G 009745-25G 009745-5G
CHMPCFF919.EA 0 EUR
CHMPCFF919. FLUO009745-1G FLUO009745-25G FLUO009745-5G
4-Chlor-3-fluoranilin
4-Chlor-3-fluoranilin
Formel: C₆H₅ClFN
MDL: MFCD01090987
CAS-Nummer: 367-22-6
EINECS: 206-683-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR