4-Chlor-2-fluor-1-nitrobenzol

Lieferant: FLUOROCHEM

Synonyme: 4-Chloro-2-fluoronitrobenzene

008545-5G 008545-25G
FLUO008545-5GEA 97.2 EUR
FLUO008545-5G FLUO008545-25G
4-Chlor-2-fluor-1-nitrobenzol
4-Chlor-2-fluor-1-nitrobenzol
Formel: C₆H₃ClFNO₂
Molecular Weight: 175.55 g/mol
MDL: MFCD00042211
CAS-Nummer: 700-37-8
EINECS: 211-842-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR